0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
Sulphuric acid
Sulphuric acid 0.9 mol/l (1.8 N) in aqueous solution
Sulphuric acid 0.9 mol/l (1.8 N) in aqueous solution
FFCHFF63529020
:
FF CHEMICALS
:
Sulphuric acid 0.9 mol/l (1.8 N) in aqueous solution
FFCHFF63529020
:
FF CHEMICALS
:
Pk:
20 L
Seq:
5302
:
:
98.08 g/mol
:
Ambient
:
231-639-5
:
2796
:
8,II
:
GHS07
:
7664-93-9
:
H₂SO₄
:
MFCD00064589