0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tubing
Transfer Tubing
Tubing, PE
Tubing, PE
:
DEUTSCH NEUMANN
Tubing, PE
:
DEUTSCH NEUMANN
PE, translucent
Odorless and tasteless
Resistant to acids, bases, alcohols, and salt solutions up to temperature of 80 °C
Flexible