Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Head Protection
Safety Helmets
Accessories for helmets Airwing
Accessories for helmets Airwing
Accessories for helmets Airwing
Catalog # UVEX9790.007
Supplier:
UVEX
Expand image
Accessories for helmets Airwing
Catalog # UVEX9790.007
Supplier:
UVEX
Specifications
Colour:
Black
Description:
Forked textile chinstrap with snap fasteners and plastic eyelets
Seq:
01
Pk:
1
Specifications
About this item
Product Details
Product Details