Dry box gloves, neoprene, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
These neoprene gauntlets protect the user from penetration by liquid, gas or solid substances.
- Seamless one-piece construction
- Available as standard (non bonded neoprene) or economy version (bonded neoprene)
- Requires clamping rings to hold gloves in place
The gloves are compatible with the Scienceware® glove box BELAH50025-0000.