Order Entry
Sweden
ContactUsLinkComponent
Methyl Parathion, certified
Methyl Parathion, certified
Catalog # EHERC15890000
Supplier:  EHRENSTORFER
CAS Number:  
Methyl Parathion, certified
Catalog # EHERC15890000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :263.21 g/mol
Storage Temperature:Freezer
EINECS:206-050-1
UN:2811
ADR:6.1,I
 :GHS06
 :GHS08
 :GHS09
CAS Number:298-00-0
Formula:C₈H₁₀NO₅PS
MDL Number:MFCD00041813