Chlormephos (S-Chloromethyl O,O-diethyl phosphorodithioate)
Catalog # EHERC11330000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:234.71 g/mol
EINECS:246-538-1
CAS Number:24934-91-6
Formula:C₅H₁₂ClO₂PS₂