Chlormephos (S-Chloromethyl O,O-diethyl phosphorodithioate)
Catalog # ACCUP-329N
Supplier: AccuStandard
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
:234.71 g/mol
EINECS:246-538-1
CAS Number:24934-91-6
Formula:C₅H₁₂ClO₂PS₂