Dichlorvos (2,2-dichlorovinyl dimethyl phosphate, DDVP)
Catalog # ACCUP-036N
Supplier: AccuStandard
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
:220.98 g/mol
EINECS:200-547-7
UN:2810
ADR:6.1,II
CAS Number:62-73-7
Formula:C₄H₇Cl₂O₄P
MDL Number:MFCD00036123