0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
Dinoseb methyl ether
Dinoseb methyl ether
Dinoseb methyl ether
Dinoseb methyl ether
Catalog # EHERC12811500
Supplier:
EHRENSTORFER
CAS Number:
Expand image
Dinoseb methyl ether
Catalog # EHERC12811500
Supplier:
EHRENSTORFER
CAS Number:
Specifications
Pack type:
Glass bottle for solids
Pk:
0,1 g
Seq:
3020
Specifications
About this item
Product Details
:
:
254.24 g/mol
CAS Number:
6099-79-2
Formula:
C₁₁H₁₄N₂O₅
Product Details