Order Entry
Sweden
ContactUsLinkComponent
(2-Naphthoxy)acetic acid
(2-Naphthoxy)acetic acid
Catalog # EHERC15439000
Supplier:  EHRENSTORFER
CAS Number:  
(2-Naphthoxy)acetic acid
Catalog # EHERC15439000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :202.21 g/mol
EINECS:204-380-0
CAS Number:120-23-0
Formula:C₁₂H₁₀O₃
MDL Number:MFCD00004066