Order Entry
Sweden
ContactUsLinkComponent
Pesticide reference standard, Terbufos (S-tert-Butylthiomethyl O,O-diethylphosphorodithioate) 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Pesticide reference standard, Terbufos (S-tert-Butylthiomethyl O,O-diethylphosphorodithioate) 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL17270000AL
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, Terbufos (S-tert-Butylthiomethyl O,O-diethylphosphorodithioate) 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL17270000AL
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    13071-79-9
  • Description:
    Terbufos (S-tert-Butylthiomethyl O,O-diethylphosphorodithioate) 10 µg/ml in acetonitrile
  • Formula:
    C₉H₂₁O₂PS₃
  • Description:
    Terbufos (S-tert-Butylthiomethyl O,O-diethylphosphorodithioate)

Specifications

About this item