Pesticide reference standard, Cadusafos 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL10934000AL
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:95465-99-9
- Description:Cadusafos 10 µg/ml in acetonitrile
- Formula:C₁₀H₂₃O₂PS₂
- Description:Cadusafos