Phenylarsine oxide, Calbiochem®
Catalog # 521000-250
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:168.02 g/mol
EINECS:211-275-3
UN:3465
ADR:6.1,III
CAS Number:637-03-6
Formula:C₆H₅AsO
MDL Number:MFCD00001990