
Purpurin(1,2,4-trihydroxy-9,10-anthraquinone)
:81-54-9
:
:C₁₄H₈O₅
:MFCD00001203
:253…256 °C
:113 °C
:256.21 g/mol
:201-359-8
:GHS07
:Ambient
- Appearance:Dark red to orange-brown to brown Powder
- Infrared spectrum:Conforms
- Solubility:(1 mg/ml in NH4OH) Red to very dark red to purple
- HPLC:≥85 %
- UV Lambda max:470 to 490 nm (10 mg/l methanol)
Frequently Bought Together