4,4'-(1-Methylpropylidene)diphenol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:77-40-7
:
Formula:C₁₆H₁₈O₂
MDL Number:MFCD00020179
:242.32 g/mol
Merck Index:14,01298
Boiling Pt:412 °C (1013 hPa)
Melting Pt:124…125 °C
EINECS:201-025-1
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:125.0 to 128.0 deg-C
- Solubility in Methanol:almost transparency