Saponin, Reagent Grade
Catalog # 0163-100G
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:HDPE bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Ash:≥10%
- Loss on Drying:≥10%
- pH (20%, Water) @25 °C:4.5 – 5.5
- Solubility (20%, Water):Pass
Specification Test Results
About this item
:
Density:1.015…1.02 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:232-462-6
CAS Number:8047-15-2
MDL Number:MFCD00081981