Order Entry
Sweden
ContactUsLinkComponent
 
Corning® Bovine Pituitary Extract (BPE), Corning
  734-1311
 :  Corning
   Low Manufacturing Impact
 :  
Corning® Bovine Pituitary Extract (BPE), Corning
  734-1311
 :  Corning
   Low Manufacturing Impact
 :  

 

  • Pk:
    15 mg
  • Source:
    Pituitary glands
  • Species:
    Bovine
  • Description:
    Bovine Pituitary Extract
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO 14001 - Bedford
  • Formulation:
    Lyophilised from NaCl-containing buffer; reconstitute in serum-free medium.
  • Quality control:
    Tested for ability to promote proliferation of Foetal Bovine Heart Endothelial Cells (FBHEC) in medium containing 10% newborn calf serum
    Filtered (0,2 µm membrane) and tested and found negative for bacteria, fungi and mycoplasma
  • Recommended concentration:
    25 - 100 µg/ml medium depending on cell type.  
  • Storage and stability:
    Stable for at least three months at 2 to 8 °C (lyophilised) or three months at −20 °C (solubilised).
    Avoid multiple freeze-thaw cycles.

 

 

BPE is a broadly used supplement to culture a variety of epithelial and endothelial cells.

BPE contains growth factors (e.g., basic FGF) and hormones.

Tested for ability to promote proliferation of Fetal Bovine Heart Endothelial Cells (FBHEC) in medium containing 10% newborn calf serum. Filtered (0.2 µm membrane) and tested and found negative for the presence of bacteria, fungi, and mycoplasma.

BPE contains growth factors (e.g., basic FGF) and hormones.