0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
Density Meters
Accessories for Densito 30PX
Accessories for Densito 30PX
:
Mettler - Toledo
Accessories for Densito 30PX
:
Mettler - Toledo
Catalog No:
614-1592
635-0117
Measurement range:
0…2 g/cm³
0…2 g/cm³