Order Entry
Sweden
ContactUsLinkComponent
 
VWR®, Bovine serum albumin (BSA)
  422351S
 :  
undefined
VWR®, Bovine serum albumin (BSA)
  422351S
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    100 g
  • CAS No.:
    9048-46-8
    90604-29-8
  • Source:
    Serum
  • Species:
    Bovine
  • Storage conditions:
    Refrigerator
  • Protein synonyms:
    BSA|Fraction V|Cohn Fraction V
  • Protein/peptide name:
    Bovine Serum Albumin
  • Purity:
    98%
  • Colour:
    light yellow
  • pH:
    6.7-7.2
  • Grade:
    Molecular Biology Grade

 

 

A higher purity product than standard grade or Cohn fraction V material. It is negative for globulins that interfere in many assays.

  • Moisture: 4,5%
  • Ashes: 1,5%