Order Entry
Sweden
Orders LinkContactUsLinkComponent
Transport carriers, Mono/TripleBox
Supplier:   DELTA T
Transport carriers, Mono/TripleBox
Supplier:  DELTA T
Certificates
Transport carriers, Mono/TripleBox
Transport carriers, Mono/TripleBox
6 Options Available Below
Product Details & Documents
These extra strong insulated boxes can be used for delivery. Made from Neopor® the boxes consist of 2 different parts which can be fitted together to create different volumes.
  • Eliminates the cost of expensive refrigeration transportation
  • Maintains the required temperature
  • Reusable
The MonoTripleBox can be used to transport test tubes or other products.
Product Family Options
Product Information
  • W×D×H
    Availability
    Price
  • 478-9928
    Monobox with one Tempshell -30 °C
    Monobox with one Tempshell -30 °C
    390×290×230 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 4 975,00 kr
    4 975,00 kr
    1 * 1 Items
       4 975,00 kr
  • 478-9929
    Monobox with one Tempshell 4 °C
    Monobox with one Tempshell 4 °C
    390×290×230 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 3 400,00 kr
    3 400,00 kr
    1 * 1 Items
       3 400,00 kr
  • 478-9930
    Monobox with one Tempshell 22 °C
    Monobox with one Tempshell 22 °C
    390×290×230 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 4 375,00 kr
    4 375,00 kr
    1 * 1 Items
       4 375,00 kr
  • 478-9934
    Two parts-TripleBox with one Tempshell -30 °C and two Tempframes
    Two parts-TripleBox with one Tempshell -30 °C and two Tempframes
    390×290×350 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 10 850,00 kr
    10 850,00 kr
    1 * 1 Items
       10 850,00 kr
  • 478-9935
    Two parts-TripleBox with one Tempshell 4 °C and two Tempframes
    Two parts-TripleBox with one Tempshell 4 °C and two Tempframes
    390×290×350 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 10 850,00 kr
    10 850,00 kr
    1 * 1 Items
       10 850,00 kr
  • 478-9936
    Two parts- TripleBox with one Tempshell 22 °C and two Tempframes
    Two parts- TripleBox with one Tempshell 22 °C and two Tempframes
    390×290×350 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 11 375,00 kr
    11 375,00 kr
    1 * 1 Items
       11 375,00 kr