1,3,5-Tribenzylhexahydro-1,3,5-triazine for synthesis
Catalog # 8.18024.0025
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:357.5 g/mol
EINECS:219-831-7
CAS Number:2547-66-2
Formula:C₂₄H₂₇N₃
MDL Number:MFCD00014599