Order Entry
Sweden
ContactUsLinkComponent
Herbicide reference standard, Atrazine 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Herbicide reference standard, Atrazine 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL10330000AL
Supplier:  EHRENSTORFER
CAS Number:  
Herbicide reference standard, Atrazine 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL10330000AL
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    1912-24-9
  • Description:
    Atrazine 10 µg/ml in acetonitrile
  • Formula:
    C₈H₁₄ClN₅
  • Description:
    Atrazine

Specifications

About this item