Order Entry
Sweden
ContactUsLinkComponent
Sodium carbonate 1.5 mol/l (3 N) in aqueous solution
Sodium carbonate 1.5 mol/l (3 N) in aqueous solution
Catalog # FFCHFF225310000
Supplier:  FF CHEMICALS
CAS Number:  
undefined
Sodium carbonate 1.5 mol/l (3 N) in aqueous solution
Catalog # FFCHFF225310000
Supplier:  FF CHEMICALS
CAS Number:  

Specifications

  • Pk:
    10 L
  • Seq:
    5214

Specifications

About this item

 :
 :105.99 g/mol
Storage Temperature:Ambient
EINECS:207-838-8
CAS Number:497-19-8
Formula:Na₂CO₃
MDL Number:MFCD00003494