Order Entry
Sweden
ContactUsLinkComponent
Pyridoxal hydrochloride, Millipore®
Pyridoxal hydrochloride, Millipore®
Catalog # 545060-25
CAS Number:  
Pyridoxal hydrochloride, Millipore®
Catalog # 545060-25
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :203.62 g/mol
EINECS:200-602-5
CAS Number:65-22-5
Formula:C₈H₁₀ClNO₃
MDL Number:MFCD00012809