0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
Citric acid
Citric acid 200 g/l in aqueous solution
Citric acid 200 g/l in aqueous solution
FFCHFF63649020
:
FF CHEMICALS
:
Citric acid 200 g/l in aqueous solution
FFCHFF63649020
:
FF CHEMICALS
:
Pk:
20 L
Seq:
5302
:
:
192.13 g/mol
:
201-069-1
:
GHS05
:
Ambient
:
77-92-9
:
HOC(COOH)(CH₂COOH)₂
:
MFCD00011669