Order Entry
Sweden
ContactUsLinkComponent
Acetic acid 0.5 mol/l (0.5 N)
Acetic acid 0.5 mol/l (0.5 N)
Catalog # FFCHFF7245000
Supplier:  FF CHEMICALS
CAS Number:  
undefined
Acetic acid 0.5 mol/l (0.5 N)
Catalog # FFCHFF7245000
Supplier:  FF CHEMICALS
CAS Number:  

Specifications

  • Pk:
    5 L
  • Seq:
    5190

Specifications

About this item

 :
 :60.05 g/mol
Storage Temperature:Ambient
EINECS:200-580-7
CAS Number:64-19-7
Formula:H₃CCOOH
MDL Number:MFCD00036152