VWR®, Autoclavable Cleanroom Goggles
Katalog # 111-0614
Leverantör: VWR Collection
Specifikationer
- Type:Anti-scratch
- Colour:Blue
- Lenses:Clear PC
- Pk:1
Specifikationer
Om denna produkt
Lightweight cleanroom goggles.
- Soft, thermoplastic body with toughened polycarbonate lens
- Strap with polypropylene strap hooks
- Indirect base ventilation
- Autoclavable at 121 °C
Suitable for ISO Class 4 environments.
Certifications: EN 166:2001; Optical Class 1.
Packaging: Packed in a NEBB Certified ISO Class 4 (Class 10) cleanroom. One piece per PE bag.