Tubings, PFA, corrugated, BOLA
Made of PFA, transparent.
- Tight bending radius only causes little cross-section reduction
- Non porous and translucent
- Almost universal chemical resistance
- Highly flexible
These tubing contains circular corrugations around the longitudinal axis. It can be shortened easily by means of a tubing cutter. Applications include perfect tubing for aggressive and pure liquids or gases and good bending radius.