Order Entry
Sweden
ContactUsLinkComponent
4,4'-Bis(dimethylamino)diphenylmethane
4,4'-Bis(dimethylamino)diphenylmethane
Catalog # EHERC10651900
Supplier:  EHRENSTORFER
CAS Number:  
4,4'-Bis(dimethylamino)diphenylmethane
Catalog # EHERC10651900
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :254.38 g/mol
EINECS:202-959-2
CAS Number:101-61-1
Formula:C₁₇H₂₂N₂
MDL Number:MFCD00008317