Order Entry
Sweden
ContactUsLinkComponent
4,4'-Bis(dimethylamino)diphenylmethane 98%
4,4'-Bis(dimethylamino)diphenylmethane 98%
Certificates
undefined
4,4'-Bis(dimethylamino)diphenylmethane 98%

Specification Test Results

  • Appearance:
    White to beige to grey to brown Powder or crystals and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    86 to 92 °C
  • Titration with HClO4:
    ≥97.5 %

Specification Test Results

Related Information