Ethyl carbazate 97%
Leverantör: Thermo Fisher Scientific
Certifikat
Om denna produkt
CAS-nummer:4114-31-2
:
Formel:C₃H₈N₂O₂
MDL-nummer:MFCD00007595
:104.11 g/mol
Kokande Pt:108…110 °C (22 mmHg)
Smält Pt:44…48 °C
Flash Pt:86 °C
Förvaringstemperatur:Ambient
EINECS:223-903-3
FN:2811
ADR:6.1,III
:GHS06
:GHS08
Specifikationstestresultat
- Appearance:White to almost white (May have a pink hue) Crystals and/or chunks or fused solid
- Infrared spectrum:Conforms
- Melting point:42 to 48 °C
- GC:≥96.0 %
- Chloride (Cl):≤20 ppm
- Sulfate (SO4):≤50 ppm
- Iron (Fe):≤20 ppm
- Heavy metals (as Pb):≤10 ppm
- Insoluble matter:≤0.01 %