(±)-2-Phenoxypropionic acid
Glass bottle for solids, 100 g
Catalog # MOLE38762652-100G
Marketsource
Supplier: Molekula
(±)-2-Phenoxypropionic acid
Catalog # MOLE38762652-100G
Supplier: Molekula
Marketsource
CAS Number: 940-31-8
Jump to:
Product Details & Documents
Synonyms: 2-Phenoxypropanoic acid, 2-Phenoxypropionic acid, alpha-Methylphenoxyacetic acid
- CAS Number:940-31-8
- Formula:C₉H₁₀O₃
- MDL Number:MFCD00002643
- Molecular Weight:166.18 g/mol
- EINECS:213-370-5
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



