
2-Phenylpyridine for synthesis, Sigma-Aldrich®
Catalog # 8.40041.0025
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 ml
- Seq:2315
Specifications
Specification Test Results
- Solubility:(20 °C) practically insoluble
- Molar Mass:155.2 g/mol
- Boiling Point:269 - 270 °C (1013 hPa)
- Vapor Pressure:- 0.5 hPa (30 °C)
- Flash Point:110 °C
- Density:1.09 g/cm3 (20 °C)
Specification Test Results
About this item
:
:155.2 g/mol
Boiling Pt:269…270 °C (1013 hPa)
Melting Pt:76…77 °C
Density:1.623 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:213-763-1
UN:2810
ADR:6.1,III
:M-Clarity™ quality level = MQ200
:GHS07
CAS Number:1008-89-5
Formula:C₁₁H₉N
MDL Number:MFCD00006280