Order Entry
Sweden
ContactUsLinkComponent
Distilling links, BOLA
Katalog # BOHLB303-02
Leverantör:  Bohlender
undefined
Distilling links, BOLA
Katalog # BOHLB303-02
Leverantör:  Bohlender
Specifikationer
  • Cone size:
    29/32 NS
  • Socket size:
    29/32 NS
  • Pk:
    1
Specifikationer
Om denna produkt
PTFE, white.
  • Temperature resistance: from −200 to +250 °C
  • Universal chemical resistance
  • Autoclavable
For vertical positioning of the Liebig condenser (e.g. in a lid of a reaction vessel). Integrated special nut (Safe-Lab) for easy locking and unlocking of the ground joint.