2,5-Dibromopyridine
Catalog # MOLE66700507-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:236.89 g/mol
Boiling Pt:234…235 °C (1013 hPa)
Melting Pt:90…93 °C
EINECS:210-839-6
:GHS07
Storage Temperature:Ambient
CAS Number:624-28-2
Formula:C₅H₃Br₂N
MDL Number:MFCD00006221