Order Entry
Sweden
ContactUsLinkComponent
Industrial gloves, Polytril™ Mix
Industrial gloves, Polytril™ Mix
Catalog # DALL2232233/9
Supplier:  HONEYWELL SAFETY
undefined
Industrial gloves, Polytril™ Mix
Catalog # DALL2232233/9
Supplier:  HONEYWELL SAFETY
Specifications
  • Glove size:
    9
  • Ambidextrous / hand-specific:
    Hand-specific
  • Disposable / reusable:
    Reusable
  • Colour:
    Black
  • Length:
    255 mm
  • EN norm:
    EN 388:2016 (4232X)
  • Cuff style:
    Knit wrist
  • Liner material:
    Cotton/PA
  • Gauge:
    10
  • Finish:
    Palm and fingers coated
  • Pk:
    10 PAIR
Specifications
About this item
Black polyamide/cotton knitted glove with rough black nitrile coating on palm and fingers.
  • Tight knit gives the glove perfect support, limits the penetration of dirt and offers a good touch
  • Ventilated back optimises the glove's breathability
  • Cotton polyamide thread offers excellent comfort and provides the glove with effective properties against cuts
  • Nitrile coating at the palm and fingertips provides outstanding abrasion resistance and impermeability to oils
Suitable for the handling of heavy and sharp objects in a greasy, damp and/or dirty environment. Also ideal for general handling in building and public works, steelworks and metallurgy.
Certifications: EN 388:2016 (4232X); Cat. II