Naphthalene 99+% for scintillation
:91-20-3
:
:C₁₀H₈
:MFCD00001742
:128.17 g/mol
:12,06457
:218 °C (760 mmHg)
:79…82 °C
:0.99 g/cm³
:78 °C
:202-049-5
:1334
:4.1,III
:GHS02
:GHS07
:GHS08
:GHS09
:Ambient
- Appearance:White Powder or crystals or flakes
- Infrared spectrum:Conforms
- Melting point:79 to 82 °C
- Solubility:(5 % in methanol) Clear colorless
- GC:≥99.0 %