Silver nitrate 99-100.5% Ph. Eur.
Catalog # 83517.180
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Assay:99 - 100.5 %
- Appearance:Colourless crystals
- Identification A:Passes test
- Identification B:Passes test
- Appearance of solution:Passes test
- Acidity or alkalinity:Passes test
- Foreign salts:Max. 0.3 %
- Aluminium, lead, copper and bismuth:Passes test
- Residual solvents:Passes test
Specification Test Results
About this item
:
:169.87 g/mol
Merck Index:12,08661
Boiling Pt:444 °C (1013 hPa)
Melting Pt:210 °C
Density:4.35 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-853-9
UN:1493
ADR:5.1,II
REACH:01-2119513705-43
:GHS03
:GHS07
:GHS05
:GHS09
CAS Number:7761-88-8
Formula:AgNO₃
MDL Number:MFCD00003414