Silver nitrate ≥99%, Proteomics Grade
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7761-88-8
:
Formula:AgNO₃
MDL Number:MFCD00003414
:169.87 g/mol
Merck Index:12,08661
Boiling Pt:444 °C (1013 hPa)
Melting Pt:210 °C
Density:4.35 g/cm³ (20 °C)
EINECS:231-853-9
UN:1493
ADR:5.1,II
REACH:01-2119513705-43
:GHS03
:GHS07
:GHS05
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Chloride:<0.0005%
- Clarity of Solution:Pass
- Copper:≤0.0002%
- Free Acid Test:Pass
- Iron:≤0.0002%
- Lead:≤0.001%
- Purity:≥99%
- Substances not Precipitated by HCl:<0.01%
- Sulphate:≤0.002%