

Ammonium nitrate ≥95% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6484-52-2
:
Formula:NH₄NO₃
MDL Number:MFCD00011425
:80.04 g/mol
Merck Index:14,534
Boiling Pt:210 °C (dec.)
Melting Pt:170 °C
Density:1.72
Storage Temperature:Ambient
EINECS:229-347-8
UN:1942
ADR:5.1,III
:GHS03
:GHS07