Magnesiumklorid, ultra dry ≥99.99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7786-30-3
:
Formula:MgCl₂
MDL Number:MFCD00011106
:95.21 g/mol
Merck Index:14,5662
Boiling Pt:1412 °C (1013 hPa)
Melting Pt:714 °C
Density:2,32
:Ampuled under argon
Storage Temperature:Omgivande
EINECS:232-094-6