Order Entry
Sweden
Orders LinkContactUsLinkComponent
Sodium thiosulphate

Sodium thiosulphate

25 kg
Catalog # SWEDAN984403
Supplier:   SWED HANDLING
Sodium thiosulphate
Catalog # SWEDAN984403
Supplier:  SWED HANDLING
CAS Number:  7772-98-7
Sodium thiosulphate

25 kg

CAS Number:  7772-98-7
Price
4 250,00 kr
Log in for availability
1 * 25 kg
Product Details & Documents
Synonyms:  Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate
  • CAS Number:7772-98-7
  • Formula:Na₂S₂O₃
  • MDL Number:MFCD00003499
  • Molecular Weight:158.11 g/mol
  • Merck Index:12,08844
  • Boiling Pt:100 °C (1013 hPa)
  • Melting Pt:48 °C
  • Density:1.66 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-867-5
Specifications

Specifications

  • Pk:
    25 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.