Order Entry
Sweden
ContactUsLinkComponent
Orthophosphoric acid ≥99%, EMSURE® for analysis, Supelco®
Corrosive
Orthophosphoric acid ≥99%, EMSURE® for analysis, Supelco®
Catalog # 1.00565.0500
Supplier:  Merck
CAS Number:  
Corrosive
Orthophosphoric acid ≥99%, EMSURE® for analysis, Supelco®
Catalog # 1.00565.0500
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Solubility:
    750 - 850 g/l (20 °C)
  • Melting Point:
    42 °C
  • Molar Mass:
    97.99 g/mol
  • Boiling Point:
    213 °C (1013 hPa)
  • Vapor Pressure:
    2 hPa (20 °C)
  • Density:
    1.87 g/cm3 (25 °C)
  • pH:
     - 0.5 (100 g/l, H2O, 20 °C)

Specification Test Results

About this item

 :
 :98 g/mol
Melting Pt:28 °C
Density:1.71…1.87 g/cm³ (25 °C)
Storage Temperature:Ambient
ADR:8,III
 :M-Clarity™ quality level = MQ300
EINECS:231-633-2
UN:3453
 :GHS05
CAS Number:7664-38-2
Formula:H₃PO₄
MDL Number:MFCD00011340