Order Entry
Sweden
ContactUsLinkComponent
Natriumhydroxid NF, Multi-Compendial, J.T.Baker®
Corrosive
Natriumhydroxid NF, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates
Corrosive
Natriumhydroxid NF, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Appearance of solution:
    passes test
  • Assay:
    97.0 - 100.5 %
  • Carbonates:
    max. 2.0 %
  • Chlorides:
    max. 200 ppm
  • Heavy Metals:
    max. 20 ppm
  • Identification A:
    passes test
  • Identification B:
    passes test
  • Iron:
    max. 10 ppm
  • Sulphates:
    max. 200 ppm
  • Chloride:
    max. 0.050 %
  • Clarity and Color of Solution:
    passes test
  • Identification 1:
    passes test
  • Identification 2:
    passes test
  • Mercury:
    passes test
  • Potassium:
    passes test
  • Sodiumcarbonate:
    max. 2.0 %
  • Identification:
    passes test
  • Insoluble substances and organic matters:
    passes test

Specification Test Results

Related Information

Frequently Bought Together