
Dilithium oxide ≥99.5% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12057-24-8
:
Formula:Li₂O
MDL Number:MFCD00016183
:29.88 g/mol
Merck Index:14,5538
Melting Pt:1427 °C
Density:2.013
Storage Temperature:Ambient
EINECS:235-019-5
UN:3262
ADR:8,II
:GHS05
Related Information
Frequently Bought Together