Order Entry
Sweden
ContactUsLinkComponent
 
Water, EMSURE® for analysis, Supelco®
 :  Merck
 
undefined
Water, EMSURE® for analysis, Supelco®
 :  Merck
 

 

  • Molar Mass:
    18.02 g/mol
  • Boiling Point:
    100 °C (1013 hPa)
  • Vapor Pressure:
    23 hPa (20 °C)
  • Density:
    1.00 g/cm3 (20 °C)
  • pH:
    (H2O, 20 °C) neutral