Potassium nitrate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7757-79-1
:
Formula:KNO₃
MDL Number:MFCD00011409
:101.1 g/mol
Merck Index:14,7648
Boiling Pt:400 °C (dec.)
Melting Pt:334 °C
Density:2.109
Storage Temperature:Ambient
EINECS:231-818-8
UN:1486
ADR:5.1,III
REACH:01-2119488224-35
:GHS03