Order Entry
Sweden
Orders LinkContactUsLinkComponent
Sodium nitrite

Sodium nitrite

25 kg
Catalog # SWEDNN51544
Oxidizing
Toxic
Environment
Supplier:   SWED HANDLING
Sodium nitrite
Catalog # SWEDNN51544
Supplier:  SWED HANDLING
CAS Number:  7632-00-0
Sodium nitrite

25 kg

CAS Number:  7632-00-0
Price
4 750,00 kr
Log in for availability
1 * 25 kg
Product Details & Documents
Synonyms:  Diazotizing salts
  • CAS Number:7632-00-0
  • Formula:NaNO₂
  • MDL Number:MFCD00011118
  • Molecular Weight:69 g/mol
  • Merck Index:13,08721
  • Boiling Pt:320 °C (1013 hPa)
  • Melting Pt:284 °C
  • Density:2.168 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-555-9
  • UN:1500
  • ADR:5.1,III
Specifications

Specifications

  • Pk:
    25 kg

Specifications