

Sodium azide ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:26628-22-8
:
Formula:NaN₃
MDL Number:MFCD00003536
:65.01 g/mol
Merck Index:13,08653
Melting Pt:275 °C
Density:1.85 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:247-852-1
UN:1687
ADR:6.1,II
:GHS06
:GHS09
Specification Test Results
- Assay:Min. 99.0 %
- Appearance:White crystalline powder
- Identification:Passes test
- Free alkalinity:Max. 0.05 meq/g
- Insolubility in water:Max. 0.015 %
- Cl (Chloride):Max. 10 ppm
- SO4 (Sulphate):Max. 50 ppm
- Cr (Chromium):Max. 20 ppm
- Cu (Copper):Max. 5 ppm
- Fe (Iron):Max. 10 ppm
- Pb (Lead):Max. 10 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number250 ml Plastic bottle for solids DIN 60 (ZPACK13250250) 6L Plastic bottle for solids DIN 80 (ZPACK13256000) 750 ml Plastic bottle for solids DIN 60 (ZPACK13250750)
Packaging Information
27417595.pdf spec_VWRC103692K spec_VWRC103694M spec_VWRC103694M spec_VWRC10369AS
Product specification
More