Order Entry
Sweden
ContactUsLinkComponent
Sodium periodate
Sodium periodate
Catalog # MOLE61500407-100G
Supplier:  Molekula
CAS Number:  
Sodium periodate
Catalog # MOLE61500407-100G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
Merck Index:13,08714
Boiling Pt:300 °C (1013 hPa)
Melting Pt:175 °C
Density:3.87 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:232-197-6
UN:1479
ADR:5.1,II
 :213.89 g/mol
 :GHS03
CAS Number:7790-28-5
Formula:NaIO₄
MDL Number:MFCD00003534